Gene Transcripts

Gene Symbol
LOC103694656
Entrez Gene ID
103694656
Gene Name
glutaredoxin-1-like
Gene Aliases
Chromosome Location
Chr. Y|NW_007906421: 3483582 - 3489147 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773748.1 XP_008771970.1 1 80

Gene Symbol
LOC103691228
Entrez Gene ID
103691228
Gene Name
glutaredoxin-1-like
Gene Aliases
Chromosome Location
Chr. 1: 179217628 - 179223199 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759849.1 XP_008758071.1 1 80

View Full Product Details