Gene Transcripts

Gene Symbol
CLEC2B
Entrez Gene ID
9976
Gene Name
C-type lectin domain family 2 member B
Gene Aliases
AICL, CLECSF2, HP10085, IFNRG1
Chromosome Location
Chr. 12: 9852369 - 9869859 on Build GRCh38
UniGene ID
Hs.85201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005127.2 NP_005118.2 3 845
GenBank mRNA AB015628.1 - 3 338
AK307334.1 - 3 372
AK311232.1 - 2 265
AK313916.1 - 3 349
AY142147.1 - 3 322
BC005254.1 - 3 339 3996700
CB160326.1 - 1 68
X96719.1 - 3 320

View Full Product Details