Gene Transcripts
- Gene Symbol
- SNCG
- Entrez Gene ID
- 6623
- Gene Name
- synuclein gamma
- Gene Aliases
- BCSG1, SR
- Chromosome Location
- Chr. 10: 86958531 - 86963260 on Build GRCh38
- UniGene ID
- Hs.349470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003087.2 |
NP_003078.2
|
1, 2 |
285 |
|
XM_011540105.2 |
XP_011538407.1
|
1, 2 |
228 |
|
GenBank mRNA |
AF010126.1 |
- |
1, 2 |
129 |
|
AF017256.1 |
- |
1, 2 |
166 |
|
AF411524.1 |
- |
1, 2 |
129 |
|
AK223367.1 |
- |
1, 2 |
242 |
|
AK312141.1 |
- |
1, 2 |
168 |
|
BC014098.2 |
- |
1, 2 |
188 |
4546444 |
BP346786.1 |
- |
1, 2 |
380 |
|
CR541788.1 |
- |
1, 2 |
118 |
|
CR541790.1 |
- |
1, 2 |
118 |
|
View Full Product Details