Gene Transcripts

Gene Symbol
SSX3
Entrez Gene ID
10214
Gene Name
SSX family member 3
Gene Aliases
CT5.3
Chromosome Location
Chr. X: 48346428 - 48356753 on Build GRCh38
UniGene ID
Hs.558445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021014.3 NP_066294.1 6 505
XM_011543885.2 XP_011542187.1 6 458
GenBank mRNA AK310513.1 - 6 458
BC005904.1 - 6 459 4064072
BC103862.1 - 6 504 40003336
S82471.1 - 5 365
U90840.1 - 6 418

View Full Product Details