Gene Transcripts
- Gene Symbol
- KCNK4
- Entrez Gene ID
- 50801
- Gene Name
- potassium two pore domain channel subfamily K member 4
- Gene Aliases
- K2p4.1, TRAAK, TRAAK1
- Chromosome Location
- Chr. 11: 64291302 - 64300031 on Build GRCh38
- UniGene ID
- Hs.647233
- Species
- Homo sapiens
- Gene Symbol
- KCNK4-TEX40
- Entrez Gene ID
- 106780802
- Gene Name
- KCNK4-TEX40 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 11: 64291722 - 64304769 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_133662.1 |
|
5 |
785 |
|
View Full Product Details