Gene Transcripts

Gene Symbol
LTB4R2
Entrez Gene ID
56413
Gene Name
leukotriene B4 receptor 2
Gene Aliases
BLT2, BLTR2, JULF2, KPG_004, LTB4-R 2, LTB4-R2, NOP9
Chromosome Location
Chr. 14: 24310151 - 24312053 on Build GRCh38
UniGene ID
Hs.642693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164692.2 NP_001158164.1 2 1018
NM_019839.4 NP_062813.2 2 962
GenBank mRNA AB029892.1 - 1 847
AB032558.1 - 2 928
AB044402.1 - 1 847
AF277230.1 - 1 2558
AF308571.1 - 1 847
BC038437.1 - 2 998

Gene Symbol
LTB4R
Entrez Gene ID
1241
Gene Name
leukotriene B4 receptor
Gene Aliases
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7
Chromosome Location
Chr. 14: 24311499 - 24318036 on Build GRCh38
UniGene ID
Hs.567248
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143919.2 NP_001137391.1 1 13
GenBank mRNA D89079.1 - 1 13

View Full Product Details