Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005692.4 | NP_005683.2 | 13 | 1752 | |
NM_007189.2 | NP_009120.1 | 13 | 1752 | ||
XM_017011639.1 | XP_016867128.1 | 13 | 1629 | ||
XM_017011640.1 | XP_016867129.1 | 13 | 1714 | ||
GenBank mRNA | AF261091.1 | - | 13 | 1609 | |
AF261092.1 | - | 13 | 1590 | ||
AJ005016.1 | - | 11 | 1279 | ||
AL050291.1 | - | 12 | 1544 | ||
BC001661.2 | - | 13 | 1584 | 2988125 | |
BT007451.1 | - | 12 | 1498 |