Gene Transcripts

Gene Symbol
NKX2-6
Entrez Gene ID
137814
Gene Name
NK2 homeobox 6
Gene Aliases
CSX2, CTHM, NKX2F, NKX4-2
Chromosome Location
Chr. 8: 23702451 - 23706598 on Build GRCh38
UniGene ID
Hs.532654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136271.2 NP_001129743.2 2 548
GenBank mRNA CN272646.1 - 2 359

View Full Product Details