Gene Transcripts

Gene Symbol
MYL6B
Entrez Gene ID
140465
Gene Name
myosin light chain 6B
Gene Aliases
MLC1SA
Chromosome Location
Chr. 12: 56152420 - 56157987 on Build GRCh38
UniGene ID
Hs.632731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199629.1 NP_001186558.1 2, 3 503
NM_002475.4 NP_002466.1 2, 3 439
GenBank mRNA AK304594.1 - 2, 3 272
AK310996.1 - 2, 3 269
BC012425.1 - 1, 2 227 3862144
BC014400.1 - 2, 3 503 3503030
BU166567.1 - 326
M31211.1 - 1, 2 207
X16434.1 - 1, 2 192

View Full Product Details