Gene Transcripts

Gene Symbol
LDLRAD2
Entrez Gene ID
401944
Gene Name
low density lipoprotein receptor class A domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 21812209 - 21825221 on Build GRCh38
UniGene ID
Hs.745158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013693.2 NP_001013715.2 4 977
XM_005245873.4 XP_005245930.1 4 1033
GenBank mRNA AK127275.1 - 4 977
BC146860.1 - 4 879 9056788

View Full Product Details