Gene Transcripts

Gene Symbol
C16orf74
Entrez Gene ID
404550
Gene Name
chromosome 16 open reading frame 74
Gene Aliases
Chromosome Location
Chr. 16: 85707517 - 85751083 on Build GRCh38
UniGene ID
Hs.461655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206967.2 NP_996850.1 4 390
GenBank mRNA AB115764.1 - 4 376
AB115765.1 - 4 388
AB115766.1 - 2 998
CD674248.1 - 4 390

View Full Product Details