Gene Transcripts

Gene Symbol
FAM25A
Entrez Gene ID
643161
Gene Name
family with sequence similarity 25 member A
Gene Aliases
bA96C23.5
Chromosome Location
Chr. 10: 87020289 - 87024730 on Build GRCh38
UniGene ID
Hs.729945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146157.2 NP_001139629.1 2 124
GenBank mRNA BC171915.1 - 2 114

View Full Product Details