Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198838.1 | NP_001185767.1 | 3 | 3005 | |
NM_001198840.1 | NP_001185769.1 | 2 | 2962 | ||
NM_006047.5 | NP_006038.2 | 3 | 3047 | ||
NM_152838.3 | NP_690051.1 | 3 | 3015 | ||
GenBank mRNA | AB015336.1 | - | 1 | 1075 | |
AB018308.2 | - | 2 | 2894 | ||
AF345332.1 | - | 3 | 2971 | ||
AF393214.1 | - | 3 | 2939 | ||
AJ289772.1 | - | 2 | 2821 | ||
AK026963.1 | - | 1 | 2405 | ||
AK074945.1 | - | 1 | 1780 | ||
AK092239.1 | - | 3 | 2959 | ||
AK299371.1 | - | 4 | 2545 | ||
AL833222.1 | - | 2 | 2809 | ||
AL834472.1 | - | 1 | 1937 | ||
BC012787.2 | - | 3 | 2995 | 3956772 | |
BC013981.2 | - | 3 | 2995 | 3845090 |