Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Chromosome Location
Chr. 17: 81977550 - 82018929 on Build GRCh38
UniGene ID
Hs.298351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251888.1 NP_001238817.1 16 1915
NM_024083.3 NP_076988.1 15 1633
NR_045351.1 14 1577
XM_011523601.2 XP_011521903.1 15 1633
XM_011523602.2 XP_011521904.1 15 1633
XM_017025036.1 XP_016880525.1 15 1708
XM_017025037.1 XP_016880526.1 15 1708
XM_017025038.1 XP_016880527.1 16 1719
XM_017025039.1 XP_016880528.1 16 1636
XM_017025040.1 XP_016880529.1 15 1573
XM_017025041.1 XP_016880530.1 14 1576
XM_017025042.1 XP_016880531.1 15 1708
XM_017025043.1 XP_016880532.1 14 1576
XM_017025044.1 XP_016880533.1 16 1644
GenBank mRNA AF324219.2 - 15 1558
AK057403.1 - 16 1915
AK057851.1 - 14 1560
AK090682.1 - 7 3069
AK126967.1 - 8 5293
AK127159.1 - 8 2205
AK127311.1 - 1 3770
AK226086.1 - 15 1506
AK290624.1 - 15 1549
BC006152.1 - 16 1708 3357489
BC018722.1 - 15 1538 4640076

View Full Product Details