Gene Transcripts

Gene Symbol
TENM1
Entrez Gene ID
10178
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, ODZ3, TEN-M1, TNM, TNM1
Chromosome Location
Chr. X: 124374680 - 125204305 on Build GRCh38
UniGene ID
Hs.23796
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163278.1 NP_001156750.1 25 5013
NM_001163279.1 NP_001156751.1 25 5010
NM_014253.3 NP_055068.2 24 4992
XM_011531230.2 XP_011529532.1 28 5654
XM_011531236.2 XP_011529538.2 14 4231
XM_011531237.2 XP_011529539.1 4 1115
XM_017029208.1 XP_016884697.1 29 5753
XM_017029209.1 XP_016884698.1 30 5808
XM_017029210.1 XP_016884699.1 28 5681
XM_017029211.1 XP_016884700.1 29 5759
XM_017029212.1 XP_016884701.1 29 5370
XM_017029213.1 XP_016884702.1 28 5682
XM_017029214.1 XP_016884703.1 27 5633
GenBank mRNA AF100772.1 - 24 4992
BC140783.1 - 25 5010 9021651
BC144543.1 - 25 5007 9053070

View Full Product Details