Gene Transcripts

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
Chromosome Location
Chr. 3: 37862152 - 37984469 on Build GRCh38
UniGene ID
Hs.475963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008392.1 NP_001008393.1 4, 5 385
NM_005808.2 NP_005799.2 3, 4 352
XM_017005519.1 XP_016861008.1 4, 5 411
XM_017005520.1 XP_016861009.1 4, 5 1729
GenBank mRNA AI143812.1 - 4, 5 385
AI192993.1 - 3, 4 352
AJ575644.1 - 3, 4 352
AJ575645.1 - 4, 5 385
AY279532.1 - 3, 4 330
AY364238.1 - 3, 4 330
D88153.1 - 4, 5 702

View Full Product Details