Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011510199.1 | XP_011508501.1 | 2 | 1194 | |
XM_011510200.1 | XP_011508502.1 | 3 | 214 | ||
XM_011510201.1 | XP_011508503.1 | 4 | 400 | ||
XM_011510202.1 | XP_011508504.1 | 2 | 139 | ||
XM_017002956.1 | XP_016858445.1 | 3 | 1448 | ||
XM_017002957.1 | XP_016858446.1 | 3 | 1300 | ||
XM_017002958.1 | XP_016858447.1 | 3 | 1287 | ||
XM_017002959.1 | XP_016858448.1 | 4 | 360 | ||
XM_017002960.1 | XP_016858449.1 | 3 | 1448 | ||
XM_017002961.1 | XP_016858450.1 | 2 | 1194 | ||
XM_017002962.1 | XP_016858451.1 | 3 | 214 | ||
XM_017002964.1 | XP_016858453.1 | 2 | 1194 | ||
XM_017002965.1 | XP_016858454.1 | 1 | 523 | ||
GenBank mRNA | BX648271.1 | - | 1 | 2934 | |
BX649148.1 | - | 1 | 3736 | ||
CR627439.1 | - | 2 | 1145 | ||
CR627446.1 | - | 1 | 3246 |