Gene Transcripts

Gene Symbol
Psg28
Entrez Gene ID
114871
Gene Name
pregnancy-specific glycoprotein 28
Gene Aliases
cea16
Chromosome Location
Chr. 7: 18422536 - 18432055 on Build GRCm38
UniGene ID
Mm.362025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054063.4 NP_473404.3 5 1845
GenBank mRNA AF113598.1 - 5 1695
BC099402.1 - 5 1831 30787049

View Full Product Details