Gene Transcripts

Gene Symbol
Klra9
Entrez Gene ID
16640
Gene Name
killer cell lectin-like receptor subfamily A, member 9
Gene Aliases
LY49I1, Ly49I
Chromosome Location
Chr. 6: 130178626 - 130194374 on Build GRCm38
UniGene ID
Mm.457996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010651.3 NP_034781.2 4 441
GenBank mRNA AY860970.1 - 3 278
U34891.1 - 3 278
U49865.1 - 441
U49866.1 - 441

Gene Symbol
Klra19
Entrez Gene ID
93971
Gene Name
killer cell lectin-like receptor, subfamily A, member 19
Gene Aliases
Ly49s
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.14112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053154.2 NP_444384.1 4 382
GenBank mRNA AF288378.1 - 4 382
BC116822.1 - 4 341 40126141
BC116824.1 - 4 335 40126143

Gene Symbol
Klra3
Entrez Gene ID
16634
Gene Name
killer cell lectin-like receptor, subfamily A, member 3
Gene Aliases
5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
Chromosome Location
Chr. 6: 130323289 - 130337626 on Build GRCm38
UniGene ID
Mm.425338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC099867.1 - 284
BC099878.1 - 284 40040424
BC099879.1 - 284 40040423
BC099880.1 - 284 40040422
BC101952.1 - 4 328 40044699
BC103543.1 - 4 328 40044703
BC103544.1 - 4 362 40044701
BC103545.1 - 4 328 40044700
BC132524.1 - 4 333 40130801
BC132526.1 - 4 333 40130803
BC145192.1 - 4 333 9053743
BC145193.1 - 4 367 9053744
U09739.1 - 4 452
U10305.1 - 4 368
U49868.1 - 4 433
U56405.1 - 4 327

Gene Symbol
Klra6
Entrez Gene ID
16637
Gene Name
killer cell lectin-like receptor, subfamily A, member 6
Gene Aliases
Ly49f, ly-49f
Chromosome Location
Chr. 6: 130013033 - 130026960 on Build GRCm38
UniGene ID
Mm.14112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008464.2 NP_032490.1 4 358
GenBank mRNA U10092.1 - 4 358

View Full Product Details