Gene Transcripts

Gene Symbol
Pex16
Entrez Gene ID
18633
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
Chromosome Location
Chr. 2: 92374676 - 92381220 on Build GRCm38
UniGene ID
Mm.202782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145122.2 NP_660104.2 7 986
GenBank mRNA AK020743.1 - 7 696
AK143073.1 - 7 988
AK162716.1 - 7 686
BC010822.1 - 7 727 4218547

View Full Product Details