Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081107.1 | NP_001074576.1 | 9 | 1846 | |
GenBank mRNA | AF436846.1 | - | 7 | 828 | |
AK052427.1 | - | 9 | 1806 | ||
BC082601.1 | - | 9 | 1810 | 30662103 | |
BC109169.2 | - | 9 | 1714 | ||
BC109170.2 | - | 9 | 1719 | 40048258 |