Gene Transcripts

Gene Symbol
Cpped1
Entrez Gene ID
223978
Gene Name
calcineurin-like phosphoesterase domain containing 1
Gene Aliases
C530044N13Rik
Chromosome Location
Chr. 16: 11803684 - 11909475 on Build GRCm38
UniGene ID
Mm.442728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146067.3 NP_666179.2 3 715
GenBank mRNA AK029379.1 - 3 639
AK032288.1 - 3 678
AK041329.1 - 3 688
AK049721.1 - 3 716
AK080876.1 - 3 673
AK133977.1 - 3 709
BC025476.1 - 3 636
BC034347.1 - 3 357 2654874

View Full Product Details