Gene Transcripts
- Gene Symbol
- Arih2
- Entrez Gene ID
- 23807
- Gene Name
- ariadne homolog 2 (Drosophila)
- Gene Aliases
- AI843547, ARI2, TRIAD1, UIP48
- Chromosome Location
- Chr. 9: 108602942 - 108654719 on Build GRCm38
- UniGene ID
- Mm.290447
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011790.4 |
NP_035920.1
|
4, 5 |
618 |
|
GenBank mRNA |
AF124664.1 |
- |
3, 4 |
445 |
|
AJ130975.1 |
- |
3, 4 |
477 |
|
AK011094.1 |
- |
4, 5 |
622 |
|
AK045012.1 |
- |
|
601 |
|
AK149936.1 |
- |
4, 5 |
598 |
|
AK165028.1 |
- |
4, 5 |
556 |
|
AK167104.1 |
- |
4, 5 |
575 |
|
AK168055.1 |
- |
4, 5 |
623 |
|
BC051998.1 |
- |
4, 5 |
618 |
6400287 |
BC052422.1 |
- |
4, 5 |
618 |
6400287 |
View Full Product Details