Gene Transcripts
- Gene Symbol
- Gm20747
- Entrez Gene ID
- 434960
- Gene Name
- predicted gene, 20747
- Gene Aliases
- Chromosome Location
- Chr. Y: 53415957 - 53418307 on Build GRCm38
- UniGene ID
- Mm.391359
- Species
- Mus musculus
- Gene Symbol
- Gm20806
- Entrez Gene ID
- 100039574
- Gene Name
- predicted gene, 20806
- Gene Aliases
- Chromosome Location
- Chr. Y: 78835721 - 78836719 on Build GRCm38
- UniGene ID
- Mm.458579
- Species
- Mus musculus
- Gene Symbol
- Gm20852
- Entrez Gene ID
- 100040786
- Gene Name
- predicted gene, 20852
- Gene Aliases
- Chromosome Location
- Chr. Y: 66739797 - 66742170 on Build GRCm38
- UniGene ID
- Mm.350930
- Species
- Mus musculus
- Gene Symbol
- Gm20854
- Entrez Gene ID
- 100040911
- Gene Name
- predicted gene, 20854
- Gene Aliases
- Chromosome Location
- Chr. Y: 85528523 - 85529519 on Build GRCm38
- UniGene ID
- Mm.457993
- Species
- Mus musculus
- Gene Symbol
- Gm20868
- Entrez Gene ID
- 100041284
- Gene Name
- predicted gene, 20868
- Gene Aliases
- Chromosome Location
- Chr. Y: 71297832 - 71300148 on Build GRCm38
- UniGene ID
- Mm.350927
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006494.1 |
- |
2 |
1025 |
|
- Gene Symbol
- Gm20917
- Entrez Gene ID
- 100042428
- Gene Name
- predicted gene, 20917
- Gene Aliases
- Chromosome Location
- Chr. Y: 79148793 - 79149787 on Build GRCm38
- UniGene ID
- Mm.458580
- Species
- Mus musculus
- Gene Symbol
- Gm21394
- Entrez Gene ID
- 100040744
- Gene Name
- predicted gene, 21394
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.464831
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC100414.1 |
- |
2 |
1035 |
|
- Gene Symbol
- Gm21943
- Entrez Gene ID
- 434935
- Gene Name
- predicted gene, 21943
- Gene Aliases
- Chromosome Location
- Chr. Y: 57531417 - 57533726 on Build GRCm38
- UniGene ID
- Mm.426195
- Species
- Mus musculus
- Gene Symbol
- Ssty2
- Entrez Gene ID
- 70009
- Gene Name
- spermiogenesis specific transcript on the Y 2
- Gene Aliases
- 1700029H17Rik, pc11
- Chromosome Location
- Chr. Y: 67045743 - 67048072 on Build GRCm38
- UniGene ID
- Mm.458581
- Species
- Mus musculus
View Full Product Details