Gene Transcripts

Gene Symbol
Rps28
Entrez Gene ID
54127
Gene Name
ribosomal protein S28
Gene Aliases
Chromosome Location
Chr. 17: 33823036 - 33824557 on Build GRCm38
UniGene ID
Mm.352374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016844.2 NP_058540.1 3 230
GenBank mRNA AK002919.1 - 3 228
AK089360.1 - 1 1282
AV497904.1 - 3 209
BC010987.1 - 3 225 4218716
BC090982.1 - 3 227 30316901
BC125644.1 - 3 294 40130159
BC125646.1 - 3 294 40130161
BY502260.1 - 3 233
U11248.1 - 3 211

View Full Product Details