Gene Transcripts

Gene Symbol
Dppa3
Entrez Gene ID
73708
Gene Name
developmental pluripotency-associated 3
Gene Aliases
2410075G02Rik, PCG7, PGC7, Stella
Chromosome Location
Chr. 6: 122626424 - 122630271 on Build GRCm38
UniGene ID
Mm.27982
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139218.1 NP_631964.1 2, 3 428
GenBank mRNA AB072734.1 - 2, 3 329
AF490347.1 - 2, 3 433
AK010723.1 - 2, 3 369
AK135769.1 - 2, 3 433
AK136263.1 - 2, 3 433
AY082485.1 - 2, 3 428
AY134859.1 - 2, 3 440
BC099433.1 - 2, 3 406 30933569
BC100331.1 - 2, 3 403 30934920
BC107340.1 - 2, 3 360 40055218
BC107341.1 - 2, 3 360 40055220

View Full Product Details