Gene Transcripts

Gene Symbol
ZER1
Entrez Gene ID
10444
Gene Name
zyg-11 related cell cycle regulator
Gene Aliases
C9orf60, ZYG, ZYG11BL
Chromosome Location
Chr. 9: 128729786 - 128772507 on Build GRCh38
UniGene ID
Hs.147950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006336.3 NP_006327.2 13 2466
XM_005251645.2 XP_005251702.1 13 2258
XM_005251646.2 XP_005251703.1 13 2455
XM_011518122.2 XP_011516424.1 13 2268
XM_017014185.1 XP_016869674.1 13 2874
XM_017014186.1 XP_016869675.1 13 2249
XM_017014187.1 XP_016869676.1 13 2868
XM_017014188.1 XP_016869677.1 13 2236
GenBank mRNA AK314911.1 - 13 2214
BC001447.2 - 2 261
BC052563.1 - 13 2451 6303965
X99802.1 - 12 2056

View Full Product Details