Gene Transcripts

Gene Symbol
Zmat5
Entrez Gene ID
67178
Gene Name
zinc finger, matrin type 5
Gene Aliases
2610510L01Rik, D11Bwg0572e, D11Bwg1548e
Chromosome Location
Chr. 11: 4704606 - 4737669 on Build GRCm38
UniGene ID
Mm.271056
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026015.2 NP_080291.1 6 733
GenBank mRNA AK007565.1 - 6 720
AK012098.1 - 6 734
AK013415.1 - 5 706
BC031796.1 - 6 712 4205598

View Full Product Details