Gene Transcripts

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
Chromosome Location
Chr. 12: 4615508 - 4649047 on Build GRCh38
UniGene ID
Hs.98397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278309.1 NP_001265238.1 5 1792
NM_006422.3 NP_006413.3 5 1770
XM_005253664.2 XP_005253721.1 5 1702
XM_011520908.1 XP_011519210.1 4 1906
XM_011520909.1 XP_011519211.1 3 1383
XM_011520910.2 XP_011519212.1 3 1838
GenBank mRNA AF087003.1 - 4 1407
AF093408.1 - 4 1474
AK074722.1 - 1 863
AK292451.1 - 5 1770
BC047535.1 - 4 1454 5267552
EU794681.1 - 4 1474
U85715.1 - 4 1447

View Full Product Details