Gene Transcripts

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location
Chr. 10: 10462515 - 11336675 on Build GRCh38
UniGene ID
Hs.309288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025076.2 NP_001020247.1 12 1533
NM_001025077.2 NP_001020248.1 12 1372
NM_001083591.1 NP_001077060.1 12 1527
NM_001326317.1 NP_001313246.1 14 1374
NM_001326318.1 NP_001313247.1 14 1392
NM_001326319.1 NP_001313248.1 16 1484
NM_001326320.1 NP_001313249.1 15 1486
NM_001326321.1 NP_001313250.1 15 1416
NM_001326323.1 NP_001313252.1 16 1528
NM_001326324.1 NP_001313253.1 13 1369
NM_001326325.1 NP_001313254.1 14 1445
NM_001326326.1 NP_001313255.1 13 1370
NM_001326327.1 NP_001313256.1 13 1388
NM_001326328.1 NP_001313257.1 14 1482
NM_001326329.1 NP_001313258.1 12 1404
NM_001326330.1 NP_001313259.1 12 1422
NM_001326331.1 NP_001313260.1 12 1574
NM_001326332.1 NP_001313261.1 12 1556
NM_001326333.1 NP_001313262.1 11 1494
NM_001326334.1 NP_001313263.1 13 1759
NM_001326335.1 NP_001313264.1 12 1568
NM_001326336.1 NP_001313265.1 13 1628
NM_001326337.1 NP_001313266.1 11 1388
NM_001326338.1 NP_001313267.1 11 1462
NM_001326339.1 NP_001313268.1 11 1480
NM_001326340.1 NP_001313269.1 13 1522
NM_001326341.1 NP_001313270.1 12 1468
NM_001326342.1 NP_001313271.1 12 1486
NM_001326343.1 NP_001313272.1 13 1540
NM_001326344.1 NP_001313273.1 12 1258
NM_001326345.1 NP_001313274.1 12 1354
NM_001326346.1 NP_001313275.1 10 1501
NM_001326347.1 NP_001313276.1 12 1678
NM_001326348.1 NP_001313277.1 12 1664
NM_001326349.1 NP_001313278.1 12 1682
NM_006561.3 NP_006552.3 12 1479
XM_005252354.4 XP_005252411.1 12 1569
XM_005252357.4 XP_005252414.1 10 1170
XM_005252358.3 XP_005252415.1 10 1511
XM_006717373.2 XP_006717436.1 11 1609
XM_011519294.2 XP_011517596.1 12 1551
XM_011519297.1 XP_011517599.1 12 1674
XM_017015542.1 XP_016871031.1 11 1257
XM_017015543.1 XP_016871032.1 12 1347
XM_017015544.1 XP_016871033.1 12 1335
XM_017015545.1 XP_016871034.1 12 1329
XM_017015546.1 XP_016871035.1 12 1329
XM_017015547.1 XP_016871036.1 12 1581
XM_017015548.1 XP_016871037.1 12 1317
XM_017015549.1 XP_016871038.1 12 1317
XM_017015550.1 XP_016871039.1 12 2315
XM_017015551.1 XP_016871040.1 12 1563
XM_017015552.1 XP_016871041.1 12 1704
XM_017015553.1 XP_016871042.1 12 1692
XM_017015554.1 XP_016871043.1 12 1674
XM_017015556.1 XP_016871045.1 11 1621
XM_017015557.1 XP_016871046.1 13 1286
XM_017015558.1 XP_016871047.1 11 1273
XM_017015559.1 XP_016871048.1 11 1609
XM_017015560.1 XP_016871049.1 11 1270
XM_017015561.1 XP_016871050.1 11 1597
XM_017015562.1 XP_016871051.1 11 1591
XM_017015563.1 XP_016871052.1 11 1578
XM_017015564.1 XP_016871053.1 10 1197
XM_017015565.1 XP_016871054.1 10 1200
XM_017015566.1 XP_016871055.1 10 1185
XM_017015567.1 XP_016871056.1 11 1325
XM_017015568.1 XP_016871057.1 13 1312
XM_017015569.1 XP_016871058.1 11 1606
XM_017015570.1 XP_016871059.1 10 1182
XM_017015571.1 XP_016871060.1 11 1594
XM_017015572.1 XP_016871061.1 11 1295
GenBank mRNA AF036956.1 - 12 1449
AF090693.1 - 12 1347
AF090694.1 - 12 1317
AF432906.1 - 12 1379
AK293164.1 - 12 1420
AK295511.1 - 10 1120
AK297313.1 - 10 1292
AK299548.1 - 12 1309
AK299992.1 - 12 1448
AK302681.1 - 9 1066
AK314087.1 - 12 1488
AK316354.1 - 12 1279
BC036391.1 - 13 1487 5260623
CR936676.1 - 1 1589
CR976278.1 - 2 120
U69546.1 - 12 1266

View Full Product Details