Gene Transcripts
- Gene Symbol
- NEK6
- Entrez Gene ID
- 10783
- Gene Name
- NIMA related kinase 6
- Gene Aliases
- SID6-1512
- Chromosome Location
- Chr. 9: 124257606 - 124352442 on Build GRCh38
- UniGene ID
- Hs.197071
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145001.2 |
NP_001138473.1
|
10, 11 |
1187 |
|
NM_001166167.1 |
NP_001159639.1
|
9, 10 |
969 |
|
NM_001166168.1 |
NP_001159640.1
|
9, 10 |
964 |
|
NM_001166169.1 |
NP_001159641.1
|
9, 10 |
973 |
|
NM_001166170.1 |
NP_001159642.1
|
9, 10 |
971 |
|
NM_001166171.1 |
NP_001159643.1
|
10, 11 |
1136 |
|
NM_014397.5 |
NP_055212.2
|
9, 10 |
1022 |
|
XM_005251664.1 |
XP_005251721.1
|
9, 10 |
1028 |
|
XM_006716936.2 |
XP_006716999.1
|
9, 10 |
1033 |
|
XM_017014217.1 |
XP_016869706.1
|
10, 11 |
1196 |
|
GenBank mRNA |
AB026289.1 |
- |
9, 10 |
953 |
|
AF087909.1 |
- |
9, 10 |
958 |
|
AK294614.1 |
- |
9, 10 |
973 |
|
AK313071.1 |
- |
9, 10 |
923 |
|
BC000101.2 |
- |
9, 10 |
968 |
3507262 |
BC004174.2 |
- |
10, 11 |
1140 |
2958695 |
BC004209.2 |
- |
10, 11 |
1140 |
2958695 |
BC012761.2 |
- |
9, 10 |
969 |
3640103 |
CR457091.1 |
- |
8, 9 |
803 |
|
CR542222.1 |
- |
8, 9 |
803 |
|
View Full Product Details