Gene Transcripts

Gene Symbol
Me1
Entrez Gene ID
17436
Gene Name
malic enzyme 1, NADP(+)-dependent, cytosolic
Gene Aliases
D9Ertd267e, Mdh-1, Mod-1, Mod1
Chromosome Location
Chr. 9: 86581363 - 86695914 on Build GRCm38
UniGene ID
Mm.148155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198933.1 NP_001185862.1 14 2049
NM_008615.2 NP_032641.2 14 1971
GenBank mRNA AK004980.1 - 14 1837
AK077968.1 - 14 1839
AK163403.1 - 14 2049
BC003287.1 - 14 1822 3582779
BC011081.1 - 14 1869 3978265
BC080660.1 - 14 1827 4240732
BC094028.1 - 14 1826 4974808

View Full Product Details