Gene Transcripts

Gene Symbol
TUBGCP2
Entrez Gene ID
10844
Gene Name
tubulin gamma complex associated protein 2
Gene Aliases
GCP-2, GCP2, Grip103, SPBC97, Spc97p, h103p, hGCP2, hSpc97
Chromosome Location
Chr. 10: 133278630 - 133312337 on Build GRCh38
UniGene ID
Hs.523370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256617.1 NP_001243546.1 18, 19 3038
NM_001256618.1 NP_001243547.1 16, 17 2765
NM_006659.3 NP_006650.1 17, 18 2954
NR_046330.1 17, 18 3301
XM_017015595.1 XP_016871084.1 10, 11 1423
XM_017015596.1 XP_016871085.1 10, 11 1423
GenBank mRNA AF042379.1 - 17, 18 2644
AK092113.1 - 17, 18 3301
AK096087.1 - 9, 10 2432
AK223586.1 - 17, 18 2842
AK297251.1 - 16, 17 2487
AK299564.1 - 17, 18 2508
AK314520.1 - 16, 17 2745
BC093770.1 - 16, 17 2633 7939615
BC111957.1 - 16, 17 2633 8327425
BC143247.1 - 17, 18 2717 9051749

View Full Product Details