Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282987.1 | NP_001269916.1 | 3 | 391 | |
NM_001282989.1 | NP_001269918.1 | 3 | 391 | ||
NM_021004.3 | NP_066284.2 | 3 | 391 | ||
XM_006720005.3 | XP_006720068.1 | 3 | 391 | ||
XM_011536366.2 | XP_011534668.1 | 3 | 391 | ||
GenBank mRNA | AB045131.1 | - | 3 | 416 | |
AF044127.1 | - | 3 | 362 | ||
AF064256.1 | - | 3 | 365 | ||
AK001870.1 | - | 3 | 357 | ||
AK301201.1 | - | 3 | 337 | ||
AK308436.1 | - | 3 | 356 | ||
AK314448.1 | - | 3 | 356 | ||
AY071856.1 | - | 3 | 342 | ||
AY358638.1 | - | 3 | 372 | ||
AY616182.1 | - | 3 | 391 | ||
BC003019.1 | - | 3 | 339 | 2822884 | |
BP366505.1 | - | 3 | 395 | ||
BU529016.1 | - | 3 | 370 | ||
DQ344810.1 | - | 3 | 370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193636.1 | NP_001180565.1 | 3 | 668 | |
NM_001193637.1 | NP_001180566.1 | 3 | 668 | ||
NM_198083.3 | NP_932349.2 | 3 | 467 | ||
GenBank mRNA | AY616183.1 | - | 603 | ||
BC000663.2 | - | 3 | 375 | 3349226 | |
DN237881.1 | - | 3 | 355 | ||
DN237882.1 | - | 4 | 409 | ||
GU564235.1 | - | 3 | 435 | ||
KC237375.1 | - | 4 | 721 | ||
KU141377.1 | - | 4 | 695 |