Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130914.1 | NP_001124386.1 | 3 | 504 | |
NM_006806.4 | NP_006797.3 | 3 | 504 | ||
XM_011529441.1 | XP_011527743.1 | 4 | 612 | ||
XM_017028265.1 | XP_016883754.1 | 4 | 642 | ||
XM_017028266.1 | XP_016883755.1 | 4 | 599 | ||
GenBank mRNA | AK289408.1 | - | 3 | 452 | |
BC011957.1 | - | 3 | 398 | 3859781 | |
BT007276.1 | - | 2 | 244 | ||
CR457061.1 | - | 2 | 244 | ||
D64110.1 | - | 3 | 338 | ||
DA381239.1 | - | 3 | 505 | ||
DR155504.1 | - | 3 | 420 |