Gene Transcripts

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Chromosome Location
Chr. 14: 52857271 - 52951192 on Build GRCh38
UniGene ID
Hs.509343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134999.1 NP_001128471.1 15 2011
NM_001135000.1 NP_001128472.1 15 2011
NM_006832.2 NP_006823.1 14 1990
XM_005267285.2 XP_005267342.1 15 2118
XM_006720008.2 XP_006720071.1 17 2163
XM_006720009.2 XP_006720072.1 16 2139
XM_006720010.2 XP_006720073.1 16 2130
GenBank mRNA AF443279.1 - 14 1885
AK126426.1 - 2 1040
AK291738.1 - 14 1943
BC017327.2 - 14 1990 4547604
BX161467.1 - 15 1932
CR749486.1 - 2 3651
EU979385.1 - 14 1825
Z24725.1 - 14 1926

View Full Product Details