Gene Transcripts

Gene Symbol
GUSBP3
Entrez Gene ID
653188
Gene Name
glucuronidase, beta pseudogene 3
Gene Aliases
Chromosome Location
Chr. 5: 69639463 - 69710527 on Build GRCh38
UniGene ID
Hs.631974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027386.2 6 912

Gene Symbol
SMA4
Entrez Gene ID
11039
Gene Name
glucuronidase beta pseudogene
Gene Aliases
SMA3, b55C20.2
Chromosome Location
Chr. 5: 70127462 - 70290177 on Build GRCh38
UniGene ID
Hs.582500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA X83300.1 - 4 653

View Full Product Details