Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
300317
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
RGD1311154
Chromosome Location
Chr. 7: 119647229 - 119677405 on Build Rnor_6.0
UniGene ID
Rn.198231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134529.1 NP_001128001.1 1 244
XM_006241976.2 XP_006242038.1 1 390
XM_006241977.2 XP_006242039.1 1 384
XM_006241978.2 XP_006242040.1 1 387
XM_006241979.2 XP_006242041.1 1 384
XM_006241981.2 XP_006242043.1 1 381
XM_006241982.2 XP_006242044.1 1 380
XM_006241983.2 XP_006242045.1 1 380
XM_008765708.1 XP_008763930.1 1 387
XM_008765709.1 XP_008763931.1 1 385
XM_008765710.1 XP_008763932.1 1 388

View Full Product Details