Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
306810
Gene Name
sushi domain containing 3
Gene Aliases
Chromosome Location
Chr. 17: 15814132 - 15830328 on Build Rnor_6.0
UniGene ID
Rn.3005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107341.1 NP_001100811.1 5 1087
XM_006253709.2 XP_006253771.1 5 2663
GenBank mRNA FQ227785.1 - 5 1272

View Full Product Details