Gene Transcripts

Gene Symbol
Hormad1
Entrez Gene ID
365868
Gene Name
HORMA domain containing 1
Gene Aliases
RGD1564960
Chromosome Location
Chr. 2: 197682000 - 197720197 on Build Rnor_6.0
UniGene ID
Rn.218539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108949.1 NP_001102419.1 9 495
XM_006232951.2 XP_006233013.1 8 450

View Full Product Details