Gene Transcripts
- Gene Symbol
- MYRF
- Entrez Gene ID
- 745
- Gene Name
- myelin regulatory factor
- Gene Aliases
- C11orf9, MRF, Ndt80, pqn-47
- Chromosome Location
- Chr. 11: 61752617 - 61788518 on Build GRCh38
- UniGene ID
- Hs.473109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127392.2 |
NP_001120864.1
|
13, 14 |
2014 |
|
NM_013279.3 |
NP_037411.1
|
13, 14 |
1914 |
|
XM_005274222.1 |
XP_005274279.1
|
13, 14 |
2002 |
|
XM_005274223.1 |
XP_005274280.1
|
13, 14 |
2002 |
|
XM_005274224.1 |
XP_005274281.1
|
13, 14 |
2002 |
|
XM_005274225.1 |
XP_005274282.1
|
13, 14 |
2002 |
|
XM_005274226.1 |
XP_005274283.1
|
13, 14 |
2002 |
|
XM_005274227.1 |
XP_005274284.1
|
13, 14 |
2002 |
|
XM_005274228.1 |
XP_005274285.1
|
13, 14 |
1849 |
|
XM_011545234.2 |
XP_011543536.1
|
13, 14 |
1888 |
|
GenBank mRNA |
AB023171.2 |
- |
13, 14 |
2002 |
|
AF038536.1 |
- |
4, 5 |
482 |
|
AF086762.1 |
- |
13, 14 |
1914 |
|
AK295013.1 |
- |
1, 2 |
243 |
|
View Full Product Details