Gene Transcripts

Gene Symbol
Stfa3l1
Entrez Gene ID
408215
Gene Name
stefin A3-like 1
Gene Aliases
Chromosome Location
Chr. 11: 67543578 - 67551211 on Build Rnor_6.0
UniGene ID
Rn.142242
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009177.1 NP_001009177.1 3 299

View Full Product Details