Gene Transcripts

Gene Symbol
Tbc1d4
Entrez Gene ID
306117
Gene Name
TBC1 domain family, member 4
Gene Aliases
RGD1561609
Chromosome Location
Chr. 15: 85927978 - 86105816 on Build Rnor_6.0
UniGene ID
Rn.233990
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770930.1 XP_008769152.1 19 4163

View Full Product Details