Gene Transcripts

Gene Symbol
MYL6B
Entrez Gene ID
140465
Gene Name
myosin light chain 6B
Gene Aliases
MLC1SA
Chromosome Location
Chr. 12: 56152420 - 56157987 on Build GRCh38
UniGene ID
Hs.632731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199629.1 NP_001186558.1 8 1023
NM_002475.4 NP_002466.1 8 959
GenBank mRNA BC012425.1 - 7 747
BC014400.1 - 8 1023 3503030
M31211.1 - 7 727
X16434.1 - 7 712

View Full Product Details