Gene Transcripts

Gene Symbol
FMOD
Entrez Gene ID
2331
Gene Name
fibromodulin
Gene Aliases
FM, SLRR2E
Chromosome Location
Chr. 1: 203340621 - 203351429 on Build GRCh38
UniGene ID
Hs.519168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002023.4 NP_002014.2 2, 3 1383
GenBank mRNA AK092899.1 - 3, 4 844
AK172740.1 - 1, 2 1082
AK291632.1 - 2, 3 1049
AK303866.1 - 3, 4 1007
BC035281.1 - 2, 3 1060 5181410
U05291.1 - 1, 2 26
X75546.1 - 1, 2 999

View Full Product Details