Gene Transcripts

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
Chromosome Location
Chr. 13: 99200425 - 99386499 on Build GRCh38
UniGene ID
Hs.508545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144072.1 NP_001137544.1 8 1311
NM_177967.3 NP_808882.1 6 1225
NR_026644.1 8 1455
XM_006719948.3 XP_006720011.1 9 1021
XM_011521082.2 XP_011519384.1 6 1081
XM_011521083.2 XP_011519385.1 7 788
XM_011521084.2 XP_011519386.1 8 876
XM_017020553.1 XP_016876042.1 3 4103
GenBank mRNA AF339812.1 - 3 295
AK054563.1 - 6 1226
AK055110.1 - 5 1008
AK075419.1 - 8 882
AK075516.1 - 6 652
AK124404.1 - 8 609
AK129861.1 - 8 910
AK290342.1 - 8 1311
AK300908.1 - 8 797
BC063559.1 - 6 670
BC072674.1 - 4 447
BC121138.1 - 6 882
BC121139.1 - 8 1054 40120987

View Full Product Details