Gene Transcripts
- Gene Symbol
- IFNAR2
- Entrez Gene ID
- 3455
- Gene Name
- interferon alpha and beta receptor subunit 2
- Gene Aliases
- IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45
- Chromosome Location
- Chr. 21: 33229895 - 33264513 on Build GRCh38
- UniGene ID
- Hs.708195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000874.4 |
NP_000865.2
|
4, 5 |
625 |
|
NM_001289125.1 |
NP_001276054.1
|
4, 5 |
625 |
|
NM_001289126.1 |
NP_001276055.1
|
4, 5 |
579 |
|
NM_001289128.1 |
NP_001276057.1
|
4, 5 |
625 |
|
NM_207584.2 |
NP_997467.1
|
4, 5 |
579 |
|
NM_207585.2 |
NP_997468.1
|
4, 5 |
579 |
|
GenBank mRNA |
AK293059.1 |
- |
3, 4 |
262 |
|
BC002793.2 |
- |
4, 5 |
573 |
3626374 |
BG715143.1 |
- |
4, 5 |
540 |
|
BI520056.1 |
- |
|
548 |
|
CD110317.1 |
- |
4, 5 |
576 |
|
CR541817.1 |
- |
3, 4 |
220 |
|
HG380509.1 |
- |
4, 5 |
345 |
|
L41942.1 |
- |
4, 5 |
546 |
|
L41943.1 |
- |
3, 4 |
259 |
|
L41944.1 |
- |
4, 5 |
546 |
|
U29584.1 |
- |
4, 5 |
286 |
|
X77722.1 |
- |
4, 5 |
445 |
|
X89772.1 |
- |
3, 4 |
220 |
|
X89814.1 |
- |
4, 5 |
384 |
|
View Full Product Details