Gene Transcripts

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
Chromosome Location
Chr. 1: 86547076 - 86580758 on Build GRCh38
UniGene ID
Hs.567422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012128.3 NP_036260.2 12 2137
NR_024602.1 11 2028
XM_011541015.2 XP_011539317.1 12 1956
GenBank mRNA AF127035.1 - 12 2121
AK000072.1 - 12 2135
AK000138.1 - 5 814
AK307785.1 - 11 1973
AK307898.1 - 10 1742
AK314430.1 - 12 2133
AY358470.1 - 12 2117
BC113687.1 - 12 2132 8322739

View Full Product Details