Gene Transcripts

Gene Symbol
GNG13
Entrez Gene ID
51764
Gene Name
G protein subunit gamma 13
Gene Aliases
G(gamma)13, h2-35
Chromosome Location
Chr. 16: 798041 - 800733 on Build GRCh38
UniGene ID
Hs.247888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016541.2 NP_057625.1 3 468
GenBank mRNA AB030207.1 - 3 447
AY029486.1 - 3 434
BM931126.1 - 1 261
DA111448.1 - 3 469

View Full Product Details