Gene Transcripts
- Gene Symbol
- PLEKHJ1
- Entrez Gene ID
- 55111
- Gene Name
- pleckstrin homology domain containing J1
- Gene Aliases
- GNRPX
- Chromosome Location
- Chr. 19: 2229952 - 2236724 on Build GRCh38
- UniGene ID
- Hs.501353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300836.1 |
NP_001287765.1
|
4, 5 |
432 |
|
NM_018049.2 |
NP_060519.1
|
4, 5 |
432 |
|
XM_017026924.1 |
XP_016882413.1
|
4 |
843 |
|
XM_017026925.1 |
XP_016882414.1
|
4 |
834 |
|
XM_017026926.1 |
XP_016882415.1
|
4, 5 |
755 |
|
XM_017026927.1 |
XP_016882416.1
|
4, 5 |
746 |
|
XM_017026928.1 |
XP_016882417.1
|
4, 5 |
746 |
|
GenBank mRNA |
AK001159.1 |
- |
4, 5 |
408 |
|
AK092742.1 |
- |
2, 3 |
1957 |
|
AK097723.1 |
- |
5, 6 |
1439 |
|
AK098725.1 |
- |
4, 5 |
522 |
|
BC003084.2 |
- |
4, 5 |
353 |
2988404 |
BC008966.2 |
- |
4, 5 |
379 |
2961278 |
CN365243.1 |
- |
4, 5 |
351 |
|
CR457253.1 |
- |
4, 5 |
327 |
|
HY085955.1 |
- |
4, 5 |
422 |
|
View Full Product Details